Avantor Seradigm Sera
The Oritain certification of product origin provides enhanced traceability and trust for Avantor® Seradigm Fetal Bovine Serum (FBS).
Request Information / Samples
Avantor® Seradigm features traceability certification from the International Serum Industry Association (ISIA) and now part of the Oritain database. Oritain's independent, scientific sampling and analysis provides added confidence for customers using Seradigm FBS. By partnering with Oritain, we are able to verify the “fingerprint” of FBS origin exclusive to the source material country of origin.  Confirmation by individual lot can be found on the product COA.
This means:
  • Enhanced traceability for regulatory compliance
  • Validation of supply chain integrity
  • Independent verification of the specified origin source on the bottle
Our proven track record for performance and consistency enables cell culture experts and other sera users to obtain the highest quality FBS for your laboratory, biopharmaceutical and other applications.
Contact your Avantor account manager today to learn more, or visit www.oritain.com/partners/ and vwr.com/seradigm.
Avantor offers a trusted supply-chain of sera with…
Quality & choice
  • Proven sera performance & consistency
    • Proprietary manufacturing methods result in lot-to-lot and bottle-to-bottle consistency
    • Quality you can see in the crystal clear, vibrant color of each bottle of sera
Expertise
  • Nutritionally superior sera
    • Unique collection & manufacturing techniques preserve more naturally-occurring growth factors found in serum
    • Industry-leading traceability & quality
    • Independent labs used for all lot release criteria
    • ISIA traceability certification confirms truth in labeling
Convenience
  • Strong supply partnerships and a vertically-integrated raw material supply chain provide long-term assurance of supply
Customization
  • Unmatched operational sera support & service
  • Flexibility in meeting each customer’s unique needs
  • Multiple lot sampling, large volume reserves, and storage solutions
  • Proven track record of operational excellence, support, and on-time delivery
What scientists are saying about Seradigm products:
We have been using Seradigm FB Essence since late 2016 and have found it to be a viable alternative to Fetal Bovine Serum (FBS). It is very cost effective and can be used to grow a variety of mammalian cancer cells, both suspension and adherent. They grow well, providing us with reproducible results - without affecting or changing the morphology of the cells. Additionally, the technical and customer service from Seradigm is excellent. I would recommend scientists to try Seradigm FB Essence – especially when grant money is very slim! – Eswar Shankar, Ph.D., Research Associate, Department of Urology, Case Western Reserve University School of Medicine
We proudly feature a fully certified supply chain for traceability by the International Serum Industry Association (ISIA). For additional information, visit www.serumindustry.org
We have established a trusted partnership with Oritain for independent origin verification testing on select FBS products. For additional information visit www.oritain.com.
Learn More
Product selection
Australia Origin FBS
Australia Origin FBS combines outstanding performance and quality with ease of regulatory access globally, suitable for a variety of applications.
Premium Grade FBS
Premium Grade FBS is widely used by cell culturists requiring high quality, high performance FBS for a variety of applications.
Ultimate Grade FBS
Ultiimate Grade FBS is the ultimate in product quality and viral safety, undergoing additional BVD virus testing using qPCR technology.
Ultra Low lgG FBS
Ultra Low lgG FBS meets a specification of ≤5 μg/mL for applications where naturally occurring levels of lgG are too high.
FB Essence
FB Essence is a nutritionally rich, cost-effective alternative to Fetal Bovine Serum (FBS) that offers pricing and supply stability without sacrificing performance.
Iron Supplemented Bovine Calf Serum (BCS)
Iron Supplemented BCS is an alternative to FBS that performs as well as FBS in many established cell lines and provides significant cost savings over FBS.