Order Entry
VWR®, Polynuclear Aromatic Hydrocarbons (PAH) Mix 3
VWR®, Polynuclear Aromatic Hydrocarbons (PAH) Mix 3
Catalog # 124832S
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR®, Polynuclear Aromatic Hydrocarbons (PAH) Mix 3
Catalog # 124832S
Supplier:  VWR Chemicals
CAS Number:  
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 1 mL
Specifications
  • Pack type:
    Glass ampoule
  • Pk:
    1 mL
  • Description:
    PAH mix in acetonitrile, 16 components
Specifications
About this item
16 components
Five μg/ml : Benzo(k)fluoranthene
10 μg/ml : Benzo(a)pyrene; Dibenz(a,h)anthracene; Fluoranthene; Benz(a)anthracene; Benzo(b)fluoranthene; Benzo(g,h,i)perylene; Indeno(1,2,3-cd)pyrene; Chrysene; Pyrene
100 μg/ml : Fluorene; Naphthalene; Acenaphthylene; Anthracene; Phenanthrene; Acenaphthene