VWR® Mr. Lab, Cleanroom Chairs and Stools
供給業者: VWR
VWR®
132-0424EA 296.43 USD
132-0424 132-0421 132-0422 132-0503
VWR® Mr. Lab, Cleanroom Chairs and Stools
椅子
All chairs and stools are available with an aluminium five star base.
  • Gas-lift seat height adjustment
  • Adjustable backrest height and inclination
  • Sealed upholstery, with special foam technology, in artificial leather covering, washable and disinfectant-proof
  • Materials suitable for use in cleanrooms
  • Smooth, sealed surfaces
  • Encapsulated mechanisms
  • Electrostatic discharge measures
Glides: The glides are wear-proof plastic, and have a large surface area, offering a high level of stability.
Castors: Dual-wheel castors, with load-sensitive brakes, have to be right for the floor. All chairs are supplied with soft castors for hard floors as standard. Hard castors for soft floors are also available as an option.
Stop&Go technology: For safety reasons, chairs whose seat is more than 650 mm high must not be movable whilst in use. With innovative Stop&Go castors, these chairs are only mobile when no one is sitting on them.
In order to create 'clean' or 'sterile' rooms, measures have to be implemented to prevent products and processes from becoming contaminated. This is a key requirement of the microelectronics sector, the pharmaceuticals industry, microsystem production, optics, medical engineering and healthcare.
証明:VWR cleanrooms chairs meet the requirements of air cleanliness Class 3 in accordance with DIN EN ISO 14644-1, cleanroom classification 1 as defined by US Federal Standard 209E, and the specifications of EU GMP guidelines. Electrostatic discharge protection according to EN 61340-5-1.
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR