VWR® Mr. Lab, 椅子、スツール
供給業者: VWR
VWR®
132-0939EA 278.77 USD
132-0939 132-0501 132-0963 132-0416 132-0966 132-0417 132-0967 132-0425 132-0964 132-0965 132-0426
VWR® Mr. Lab, 椅子、スツール
椅子
All chairs and stools are available with a five star base which is made from aluminium.
  • Chair backrest adjustable in height and inclination
  • Seat height adjustable by gas-lift
  • Aluminium five star base
The perfect combination of comfort and ergonomics for optimal performance. Anatomically-moulded cushions with pelvis and integrated lumbar support. Very comfortable, with large, soft upholstery in artificial leather covering, washable and disinfectant-proof.
Glides: The glides are wear-proof plastic and have a large surface area offering a high level of stability.
Castors: Dual-wheel castors with load-sensitive brakes have to be right for the floor. All chairs are supplied with soft castors for hard floors as standard. Hard castors for soft floors are also available as an option.
Stop&Go technology: For safety reasons chairs whose seat is more than 650 mm high must not be movable whilst in use. With innovative Stop&Go castors these chairs are only mobile when no one is sitting on them.
証明:Conform to security level S2, GMP.
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR