VWR®, ラック、エッペンチューブ用
供給業者: VWR
VWR®
211-0210EA 20.89 USD
211-0210 211-0205 211-0206 211-0204 211-0209 211-0207 211-0208
VWR®, ラック、エッペンチューブ用
ラック Tube Racks
PP.
  • For freezer use
  • Side handles for secure handling
  • Numbered for easy sample identification
Space for up to 80×1,5 or 2,0 ml microcentrifuge tubes. 5×16 array with conical-shaped holes to hold microtubes securely.
注文情報:注文情報:注文情報:The assorted colour set includes one rack each in blue, green, pink, yellow and orange.
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR