VWR® 3D Rotator Waver
Catalog # 444-0759
- Angle:0 - 20°
- Shaking motion:3-dimensional
- Speed range:1 - 30 min⁻¹*
- Description:3 dimensional rotator waver, EU-plug
- Seq:01
- Model:3-D rotator waver
- Max. load:2,3 kg**
- Platform W×D:222×299 mm
- Weight:6,8 kg
- W×D×H:279×432×152 mm
- Pk:1