VWR® ARISTAR®, Rubidium Standard Solution, 1000 mg/L Rb in dil. Nitric Acid (from RbNO3), Standard for ICP
Catalog # 455902G
Supplier: VWR Chemicals
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2-5% HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Rubidium
- Symbol:Rb
Specifications
About this item
RbNO₃ in HNO₃ 2 to 5%.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with Certificate of Analysis.
Frequently Bought Together