VWR® ARISTAR®, Lithium Standard Solution, 1000 mg/L Li, Standard for Ion Chromatography
458242F
:
- Concentration:1000 mg/L
- Matrix:H₂O/tr HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Lithium
- Symbol:Li
Li in H₂O tr.HNO₃.
Traceable to SRM from NIST, manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities.
Supplied with certificate of analysis.