ラック、冷凍ボックス用、標準タイプ
供給業者: Tenak
479-1748 479-0735 479-0736 479-1747 479-0737 479-0738 479-1749 479-0739 479-1751 479-1750 479-0740 479-1753 479-0741 479-1752 479-1755 479-1315 479-0742 479-1754 479-0743 479-0744 479-1317 479-1757 479-0745 479-1316 479-1756 479-1870 479-1715 479-1319 479-0746 479-0747 479-1758 479-1318 479-0748 479-1717 479-1716 479-0749 479-1719 479-1718 479-1003 479-1002 479-0750 479-1720 479-0751 479-1004 479-0752 479-1722 479-0753 479-0754 479-1721 479-1724 479-0755 479-0756 479-1723 479-1320 479-0757 479-1726 479-1725 479-1728 479-1727 479-1729 479-1376 479-1731 479-1730 479-1733 479-1732 479-1735 479-1734 479-1737 479-1736 479-1739 479-1738 479-1740 479-1742 479-0850 479-0851 479-1741 479-0730 479-0731 479-1744 479-0732 479-1743 479-1746 479-0733 479-1745 479-0734
ラック、冷凍ボックス用、標準タイプ
ラック Cryogenic Box Racks
Stainless steel.
  • Racks for five different box heights
  • Can be used with all freezer brands
  • Fits all standard cryoboxes
For upright freezers. These racks are an inexpensive storage system. They will fit into any freezer, and can be used with all standard cryoboxes. Measure the inside height and depth of your freezer before choosing the right rack.
Stainless steel safety spring lock
Can be used when there is a gap between the cryobox and the shelf from 1 to 6 mm
For Tenak classic and budget racks, or any other rack brand with a similar design
Withstands temperatures below −200 °C
The safety spring lock is used to prevent the cryoboxes from sliding out when pulling the rack in and out the freezer. Simply place the spring lock on the lid of the cryobox and push the cryobox in the rack. The spring will come out together with the cryobox when pulling it out.
注文情報:注文情報:注文情報:Delivered without cryoboxes.
注意:* Shorter frame for Panasonic/Sanyo freezers (479-1870).
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR