Digital Solutions
Applications
Support
About
Order Entry
VWR®, Digital Timer, One Channel
609-0184
:
VWR Collection
VWR®, Digital Timer, One Channel
609-0184
:
VWR Collection
1 * 1 items
-
+
Colour:
White
Description:
Digital timer, single channel
W×D×H:
46×12×88 mm
Accuracy:
±0,01 %
Pk:
1