VWR®, Pipette Tips

Supplier: VWR
VWR®
612-6743EA 0 USD
612-6743 612-6765 612-6766 612-6744 612-6741 612-6763 612-6764 612-6742 612-6761 612-6740 612-6762 612-6760 612-6729 612-6749 612-6727 612-6728 612-6769 612-6747 612-6726 612-6748 612-6767 612-6745 612-6746 612-6768 612-6732 612-6754 612-6733 612-6755 612-6752 612-6730 612-6753 612-6731 612-6750 612-6751 612-6738 612-6739 612-6736 612-6758 612-6737 612-6759 612-6734 612-6756 612-6757 612-6735
VWR®, Pipette Tips
Pipette Tips
These VWR pipette tips are designed for precise liquid transfer.

  • Fully autoclavable to 121 °C
  • Colour-coded racks for easy volume identification
  • Tips with PP filter are also available
  • DNase-/RNase-free and non-pyrogenic
  • Every rack or case is printed with a batch number for traceability
  • Sterile tips are sterilized by irradiation SAL 10⁻⁶ (ISO 11137)
  • Shelf Life: 3 years after month of production

Good quality consumable products for micropipettors. Preferred accessories for most brand micropipettors.

Certifications: Manufactured in a class 100 000 room environment under ISO 13485 and ISO 9001 quality management system.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

詳細確認 About VWR