VWR®, ピペットチップ、リロードシステム
供給業者: VWR
VWR®
613-1090EA 23.62 USD
613-1090 613-1081 613-1092 613-1091 613-1094 613-1072 613-1082 613-1093 613-1063 613-1096 613-1062 613-1098 613-1087 613-1064 613-1086 613-1078 613-1100 613-1077 613-1066 613-1068
VWR®, ピペットチップ、リロードシステム
ピペットチップ
Transparent PP, sterile or non sterile, autoclavable.
  • Non sterile tips are free from DNA, DNase and RNase; E-beam sterilised tips are free from DNA, DNase and RNase, pyrogens, PCR inhibitors and endotoxins
  • Robust, traceable racks are easy to grab with their ergonomic clips
  • Reload boxes including racks or empty reload boxes are also available and feature flip lock latch for one handed opening and closing, reversible lid with lip, flush fitting and click clip to stabilise the racks
注意:SC: Single channel pipette / MC: Multi channel pipette
For more compatibility information, please contact Avantor Customer Services.
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR