Pipette tips for Rainin® LTS® pipettes
Supplier: VWR, part of Avantor
VWR®
613-0583EA 39.84 USD
613-0583 613-0728 613-5431 613-0576 613-5433 613-2109 613-5434 613-0581 613-0727 613-5429 VWRU37001-608 613-0729 VWRU89079-452 613-5430 613-0579 613-5432 613-0577 613-0722 613-2110 613-0584 613-0582 613-0721 613-0580 VWRU89079-448 613-0578 613-0730 VWRU89079-436
Pipette tips for Rainin® LTS® pipettes
Pipette Tips
These VWR® pipette tips are made of clear, pre-tested PP resin and fit Rainin® LTS® pipettes.
  • Fit and eject with significantly less force than most conventional pipette tips
  • Ultra-fine points and super-clear resin for excellent viewing of sample
  • Free of RNase, DNase, endotoxins and trace metals
  • Tested for ATP
  • Available in different packaging options
  • Autoclavable
Unlike pre-sterilised pipette tips from other brands, these tips use a validated sterile process (ISO) and can be used for medical research and diagnostics.
Packaging options:
Bulk: Tips are supplied in resealable PE bags (1000 tips). These packaging options are not recommended if you plan to store the tips for long periods and if warping is a concern.
Racks (96 tips per rack): These colour coded racks, with smooth rounded edges, have heavy hook hinges that are extremely durable, and an ergonomic snap latch that won’t catch gloves. Strong enough to be autoclaved many times. The separate rack insert includes a bevelled top and extra sturdy surface for multi-channel pipetting.
Next Generation refills: Next Generation refill systems refill VWR and other brands of pipette tip racks. Their low carbon footprint makes them an ideal solution for waste reduction goals in the lab. Users don’t need gloves to refill tip racks - Next Generation refills racks without users having to touch the new tips.
Order Now
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Learn more About VWR