Pipette tips for Rainin® LTS® pipettes, low retention
Supplier: VWR, part of Avantor
VWR®
613-5787EA 85.93 USD
613-5787 613-5794 613-5793 613-5435 613-5784 613-5439 613-5437 613-5795 613-5436 613-5791 613-5789 613-5788 613-5786 613-5785 613-5790 613-5792
Pipette tips for Rainin® LTS® pipettes, low retention
Pipette Tips
These tips are made with a special resin that reduces the potential for sample retention inside the tip. They help users reduce sample loss when performing protein research. These tips fit Rainin® LTS® pipettes.
  • Fit and eject with significantly less force than most conventional pipette tips
  • Ultra-fine points and super-clear resin for excellent viewing of sample
  • Free of RNase, DNase, endotoxins and trace metals
  • Tested for ATP
  • Autoclavable
Unlike pre-sterilised pipette tips from other brands, these tips use a validated sterile process (ISO) and can be used for medical research and diagnostics.
Packaging options:
Bulk: Tips are supplied in resealable PE bags (1000 tips). These packaging options are not recommended if you plan to store the tips for long periods and if warping is a concern.
Racks (96 tips per rack): These colour coded racks, with smooth rounded edges, have heavy hook hinges that are extremely durable and an ergonomic snap latch that won’t catch gloves. Strong enough to be autoclaved many times. The separate rack insert includes a bevelled top and extra sturdy surface for multi-channel pipetting.
Next Generation refills: Next Generation refill systems refill VWR and other brands of pipette tip racks. Their low carbon footprint makes them an ideal solution for waste reduction goals in the lab. Users don’t need gloves to refill tip racks - Next Generation refills racks without users having to touch the new tips.
Order Now
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Learn more About VWR