ピペットチップ、Chem-Resin™ (ケムレジン
供給業者: VWR
VWR®
613-6480EA 38.69 USD
613-6480 613-6484 613-6450 613-6472 613-6481 613-6477 613-6487 613-6475 613-6486 613-6447 613-6478
ピペットチップ、Chem-Resin™ (ケムレジン
ピペットチップ
These tips provides a solution for working with challenging strong polar solvents or highly electronegative reagents.
  • More neutral surface tension
  • Manual tips featuring Chem-Resin™
The reduced hydrophobicity on the surface of the tip facilitates low volume draw and drip-free dispense of these classes of chemicals.
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR