Digital Solutions
Applications
Support
About
Order Entry
VWR® Probeless Electronic Thermometers
620-1722
:
VWR Collection
VWR® Probeless Electronic Thermometers
620-1722
:
VWR Collection
1 * 1 items
-
+
Range:
−10…+50 °C
Resolution:
0,1 °C
Description:
Indoor/outdoor thermometer, W×H: 100×215 mm
Seq:
02
Accuracy:
±1,0 °C
Pk:
1