VWR® Traceable®, Indoor/Outdoor Digital Thermometer with Giant Dual-Display and Calibration
620-2713
- Range:–50...70 °C; –58...158 °F
- Resolution:0,1 °C; 0,1 °F
- Description:Digital thermometer with 2 probe channels
- Accuracy:±1,0 °C; ±2,0 °F
- Cable length:3 m
- Pk:1
- Probe:3,2 mm