Order Entry
VWR® SAS Super DUO 360 Air Samplers
VWR® SAS Super DUO 360 Air Samplers
undefined
VWR® SAS Super DUO 360 Air Samplers
  • Catalog No:
  • 710-0867
  • 710-0866
  • Flow rate:
  • 180 L/min
  • 180 L/min
  • Sampling head:
  • Aluminium, stainless steel or plastic (certified ready to use)
  • Aluminium, stainless steel or plastic (certified ready to use)
  •  
    Model
    Pk
  • Air sampler using contact plates
    Air sampler using contact plates
    710-0867
    SAS Super DUO 360
    1
    1 * 1 items
    1 * 1 items
    Accessories
    Specifications
    Environmentally Preferable
    Flow rate
    180 L/min
    Sampling head
    Aluminium, stainless steel or plastic (certified ready to use)
  • Air sampler using Petri dishes
    Air sampler using Petri dishes
    710-0866
    SAS Super DUO 360
    1
    1 * 1 items
    1 * 1 items
    Accessories
    Specifications
    Environmentally Preferable
    Flow rate
    180 L/min
    Sampling head
    Aluminium, stainless steel or plastic (certified ready to use)
Aspirating Heads
  •  
    Seq
    Description
    Pk
  • 21
    Stainless steel head for standard Ø 90 mm Petri dishes (without adapter)
    1
    1 * 1 items
  •  
    Seq
    Description
    Pk
  • 16
    Aluminum aspirating head for Ø 55 mm contact plates
    1
    1 * 1 items
  •  
    Seq
    Description
    Pk
  • 18
    Aluminium head and adapter for standard Ø 90 mm Petri dishes
    1
    1 * 1 items
  •  
    Seq
    Description
    Pk
  • 20
    Stainless steel head and adapter for standard Ø 90 mm Petri dishes
    1
    1 * 1 items
  •  
    Seq
    Description
    Pk
  • 19
    Aluminium head for standard Ø 90 mm Petri dishes (without adapter)
    1
    1 * 1 items
  •  
    Seq
    Description
    Pk
  • 17
    Stainless steel aspirating head for Ø 55 mm contact plates
    1
    1 * 1 items
  •  
    Seq
    Description
    Pk
  • 01
    Sterile heads for Ø 55 mm contact plates
    40
    1 * 40 items
  •  
    Seq
    Description
    Pk
  • 02
    Sterile heads for Ø 90 mm Petri dishes
    40
    1 * 40 items