VWR®, ピペットチップ
供給業者: VWR
VWR®
613-0278EA 60.04 USD
613-0278 613-0279 613-0830 VWRU53503-790 613-0831 613-1647 613-1646 613-0272 613-0273 613-0274 613-0277 613-0300 613-0267 613-0301 613-1634 613-0302 VWRU83007-382 613-0304 VWRU83007-386 613-0260 613-0261 613-0265 613-0266 732-0505 732-0504 732-0506 613-0726 VWRU89368-996 613-0334 613-0335 613-0731 613-0732 613-0336 613-0337 613-0734 613-0339 613-0735 613-0736 613-0296 613-0298 613-0331 613-0299 613-0332 89087-530 83007-376 89087-532 736-0206 53509-132 613-2104 53509-130 613-2106 613-0723 732-0500 613-0287 732-0502 613-0288 613-0749 VWRU53509-009 613-2137 VWRU37001-528 613-0357 613-0358 613-0359 613-0239 613-0636 613-0637 613-0350 613-0230 613-2136 613-0750 613-2135 613-0738 613-0739 613-0344 613-0345 613-0340 613-0341 613-0342 613-0343 613-2150 53508-920 53508-922 613-0259 613-0252 613-0253 613-0892 82018-840 613-0638 613-2149 613-0366 613-2148 613-0246 613-0247 613-0368 613-0369 613-0248 613-0889 613-2143 613-0240 613-2145 613-0241 613-0362 613-0242 613-2144 613-0364 613-0243 613-2146
VWR®, ピペットチップ
ピペットチップ
These tips are made of high quality, 100% pure, virgin medical grade PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard.
  • Sustainable design based on virgin resins
  • A wide seal ensures tips will seal easily and consistently without leaking
  • Guaranteed free of detectable DNA, DNase/RNase, endotoxins and heavy metals
  • Tested for endotoxin (pyrogen) contamination
  • Resins are pre-tested for metal contamination
  • Graduated tips only: Graduations allow a visual check of pipetting accuracy
  • Certificates are on file by lot number for all test results
Items (pipette tips and boxes) can be autoclaved at 122 °C for a maximum of 20 minutes at 15 psi.
Macro tips (1 to 5 and 5 to 10 ml) are supplied in composite racks (fibre carton). These racks are not autoclavable. For sterile tips, please order the sterile version.
Please note: A steam cycle should be used. Always air dry these items. Do not use a dry cycle or glassware cycle as deformation can occur to plastics that are heated for extended periods.
Tip styles:
Bevel Point: Pipette tips that feature bevelled points are designed to retain less fluid and are a good choice for a multitude of procedures.
UltraFine: Pipette tips with UltraFine™ point feature a thin flexible section at the end that is non bevelled and very flexible. This helps you dispense the last tiny amount of sample from the tip.
FlexTop: These ultimate tips feature a uniquely thin and flexible top that conforms to the pipette barrel unlike any other tips. This design allows them to mount and eject from most pipettes with forces that help eliminate the potential for repetitive strain injuries.
Wide orifice: Extra wide opening allows you to pipette macrophages or sticky DNA materials. Bevelled end helps eliminate carry-over from viscous solutions.
Packaging options:
Bulk: Tips are either supplied in resealable PE bags (1000 tips) or in one large bag inside a corrugated container (10000 tips). These packaging options are not recommended if you plan to store the tips for long periods, and if warping is a concern.
Racks (96 + 192 tips per rack): These racks, with smooth rounded edges, have heavy hook hinges that are extremely durable, and an ergonomic snap-latch that won’t catch gloves. Strong enough to be autoclaved many times. The separate rack insert includes a bevelled top and extra sturdy surface for multi-channel pipetting.
Stack racks: These high quality racks need to be assembled by the user. You simply have to insert the fully racked refills into the rack. Since the tips are never touched, you do not have to you be concerned about contamination from your hands or latex gloves. These stacks are very stable so you can pipette with a multi-channel pipette directly from the stack.
Stack rack refills: These refills are a slimmed down version of the Stack racks. Neatly packed, ready for you to refill the racks you already have.
Next Generation refills: Next Generation refill systems refill VWR and other brands of pipette tip racks. Their low carbon footprint makes them an ideal solution for waste reduction goals in the lab. Users don’t need gloves to refill tip racks - Next Generation refills racks without users having to touch the new tips. The small footprint extends to the packaging, with a reclosable box that helps maximise storage space in crowded laboratories.
Individually wrapped: Tips are individually packed in thermal-sealed PE bags. The tips have been radiation sterilised.
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR