VWR®, フィルターチップ
供給業者: VWR
VWR®
732-0526EA 65.23 USD
732-0526 732-0528 16466-008 732-0570 732-1100 VWRU53510-102 732-0571 732-1102 732-0530 732-0573 732-1101 732-0532 732-0531 732-1103 46620-328 732-0534 732-0611 732-0533 732-0536 732-0535 732-1107 732-0516 732-0537 732-0515 732-0559 53510-106 VWRU89003-420 53510-014 VWRU53510-016 VWRU53510-037 732-0541 732-0540 732-0521 732-0565 732-0543 732-0520 732-0542 732-0523 732-0566 732-0544 732-0569
VWR®, フィルターチップ
ピペットチップ
All tips are made of high quality, 100% pure, virgin medical grade PP. Made in the highest quality moulds, these tips can be used whenever the work demands a higher standard. All products are tested for endotoxin (pyrogen) contamination and certified to USP results. Resins are pre-tested for metal contamination. Certificates are on file by lot number for all test results.
  • A wider seal ensures that tips will seal easily and consistently without leaking, on old as well as new pipettes
  • Guaranteed free of detectable human DNA, DNase/RNase, ATP, endotoxins and heavy metals
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR