These aerosol filter tips, with their patented filters made from high molecular weight polyethylene, offer complete protection from sample carry-over. The filter pores have an average size of 10 μm, so even radionucleotides are captured before reaching the pipette barrel. They are ideal for PCR and DNA manipulation.
- Sustainable design based on 95% renewable materials - compostable tip base from organic non petroleum resin, carton made from 100% recycled fibres, printing is soy-based vegetable ink
- Easy to use refill system - new pipette tip racks, inserts and transfer cover preventing tips wobbling during refill and keeping tips clean
- Certified human DNA-/RNase-/DNase-free
- Free from pyrogens, ATP and protease activity
Next Generation is a complete system designed with a low carbon footprint that includes the first ever use of bio-plastic (non petroleum) compostable resin in pipette tip refills. Next Generation’s small footprint extends to the package, with a reclosable box that helps maximise storage space in crowded labs.
包装:Compact, reclosable packaging with reduced overall weight.