VWR®, Filter Tips, Next Generation Refill
供給業者: VWR
VWR®
VWRU89495-360EA 490.21 USD
VWRU89495-360 732-1481 732-1480 732-1482 732-1476
VWR®, Filter Tips, Next Generation Refill
ピペットチップ
These aerosol filter tips, with their patented filters made from high molecular weight polyethylene, offer complete protection from sample carry-over. The filter pores have an average size of 10 μm, so even radionucleotides are captured before reaching the pipette barrel. They are ideal for PCR and DNA manipulation.
  • Sustainable design based on 95% renewable materials - compostable tip base from organic non petroleum resin, carton made from 100% recycled fibres, printing is soy-based vegetable ink
  • Easy to use refill system - new pipette tip racks, inserts and transfer cover preventing tips wobbling during refill and keeping tips clean
  • Certified human DNA-/RNase-/DNase-free
  • Free from pyrogens, ATP and protease activity
Next Generation is a complete system designed with a low carbon footprint that includes the first ever use of bio-plastic (non petroleum) compostable resin in pipette tip refills. Next Generation’s small footprint extends to the package, with a reclosable box that helps maximise storage space in crowded labs.
包装:Compact, reclosable packaging with reduced overall weight.
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR