VWR®, Filter Tips for Rainin® LTS® Pipettes
供給業者: VWR
VWR®
732-0801EA 73.3 USD
VWRU89201-526 732-0801 732-1483 732-1485 732-2897 732-0800 732-0799
VWR®, Filter Tips for Rainin® LTS® Pipettes
ピペットチップ
These sterile aerosol filter tips are made of clear, pre-tested PP resin and fit Rainin® LTS® pipettes.
  • Fit and eject with significantly less force than most conventional pipette tips
  • Ultra-fine points and super-clear resin for excellent viewing of sample
  • Free of RNase, DNase, endotoxins, human DNA, protease and trace metals
  • Tested for ATP - Sustainable design thanks to use of virgin resins
Unlike pre-sterilised pipette tips from other brands, these tips use a validated sterile process (ISO) and can be used for medical research and diagnostics.
Packaging options:
Racks (96 tips per rack): These colour coded racks, with smooth rounded edges, have heavy hook hinges that are extremely durable and an ergonomic snap latch that won’t catch gloves. Strong enough to be autoclaved many times. The separate rack insert includes a bevelled top and extra sturdy surface for multi-channel pipetting.
Next Generation refills: Next Generation refill systems refill VWR® and other brands of pipette tip racks. Their low carbon footprint makes them an ideal solution to waste reduction goals for the lab. Users don’t need gloves to refill tip racks - Next Generation refills racks without users having to touch the new tips.
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR