Order Entry
VWR® Media Supplements for Dehydrated Culture Media
undefined
VWR® Media Supplements for Dehydrated Culture Media
  •  
    For
    Pack type
    Pk
  • AB supplement
    AB supplement
    85860.0001
    Legionella CYE agar base (ISO) Cat. No. 84629.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • Bacillus cereus anti-microbic supplement
    Bacillus cereus anti-microbic supplement
    84732.0001
    Mannitol egg yolk polymyxin agar base - MYP (ISO) Cat. No. 84635.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • BCYE α-growth supplement
    BCYE α-growth supplement
    84726.0001
    Legionella CYE agar base (ISO) Cat. No. 84629.0500
    4 Vial
    1 * 4 Vial
    1 * 4 Vial
  • BCYE α-growth supplement without cysteine
    BCYE α-growth supplement without cysteine
    84727.0001
    Legionella CYE agar base (ISO) Cat. No. 84629.0500
    4 Vial
    1 * 4 Vial
    1 * 4 Vial
  • Bolton broth selective supplement
    Bolton broth selective supplement
    84744.0001
    Campylobacter Bolton broth base Cat. No. 84697.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • Campylobacter Bolton selective supplement, 225 ml
    Campylobacter Bolton selective supplement, 225 ml
    928580NL
    Campylobacter Bolton broth base Cat. No. 84697.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • CCD selective supplement
    CCD selective supplement
    84742.0001
    Campylobacter blood-free medium base Bolton - MCCDA Cat. No. 84695.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • CFC selective supplement
    CFC selective supplement
    84746.0001
    Pseudomonas agar base Cat. No. 84650.0500 - in compliance with ISO 13720 (meat products)
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • Chloramphenicol
    Chloramphenicol
    84729.0001
    DG18 agar base Cat. No. 84632.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • CN selective supplement
    CN selective supplement
    84745.0001
    Pseudomonas agar base Cat. No. 84650.0500 - in compliance with EN 12780
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • d-Cycloserine selective supplement, 40 mg
    d-Cycloserine selective supplement, 40 mg
    928610NL
    TSC (ISO) Cat. No. 84905.0500 and Cat. No. 85054.0500
    10
    1 * 10 items
    1 * 10 items
  • d-Cycloserine supplement
    d-Cycloserine supplement
    84734.0001
    Tryptose sulphite cycloserine (TSC) agar Cat. No. 84636.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • Egg yolk emulsion
    Egg yolk emulsion
    84733.0001
    Mannitol egg yolk polymyxin agar base - MYP (ISO) Cat. No. 84635.0500
    50 mL
    1 * 50 mL
    1 * 50 mL
  • Fraser half selective supplement
    Fraser half selective supplement
    84721.0001
    Fraser broth to prepare half Fraser broth Cat. No. 84611.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • Fraser selective supplement
    Fraser selective supplement
    84720.0001
    Fraser broth Cat. No. 84611.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • Glycerol for up to 10 L media
    Glycerol for up to 10 L media
    84730.0001
    Pseudomonas agar base Cat. No. 84650.0500; Pseudomonas selective (cetrimide) agar (Ph. Eur., USP, JP) Cat. No. 84638.0500; Wort broth Cat. No. 84660.0500; Wort agar Cat. No. 84661.0500
    100 mL
    1 * 100 mL
    1 * 100 mL
  • GVPC supplement
    GVPC supplement
    84725.0001
    Legionella CYE agar base (ISO) Cat. No. 84629.0500
    4 Vial
    1 * 4 Vial
    1 * 4 Vial
  • Iodine iodide solution for 2,5 L media in total
    Iodine iodide solution for 2,5 L media in total
    84723.0001
    Muller-Kauffmann tetrathionate-novobiocin broth (MKTTn) (ISO) Cat. No. 84624.0500
    50 mL
    1 * 50 mL
    1 * 50 mL
  • Karmali selective supplement
    Karmali selective supplement
    84743.0001
    Karmali medium base Cat. No. 84696.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • MWY selective supplement
    MWY selective supplement
    85861.0001
    Legionella CYE agar base (ISO) Cat. No. 84629.0500
    4 Vial
    1 * 4 Vial
    1 * 4 Vial
  • Novobiocin MKTTn supplement
    Novobiocin MKTTn supplement
    84722.0001
    Muller-Kauffmann tetrathionate-novobiocin broth (MKTTn) (ISO) Cat. No. 84624.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • Oxford supplement
    Oxford supplement
    84728.0001
    Oxford agar base Cat. No. 84630.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • Palcam supplement
    Palcam supplement
    84724.0001
    Palcam agar base (ISO) Cat. No. 84625.0500
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • Potassium tellurite 1% supplement
    Potassium tellurite 1% supplement
    900010ZK
    Giolitti-Cantoni broth agar base (ISO)
    100 mL
    1 * 100 mL
    1 * 100 mL
  • PP selective supplement
    PP selective supplement
    84747.0001
    Pseudomonas agar base Cat. No. 84650.0500 - in compliance with ISO 11059 (milk and dairy products)
    10 Vial
    1 * 10 Vial
    1 * 10 Vial
  • TTC 1% solution for 3 L media in total
    TTC 1% solution for 3 L media in total
    84731.0001
    KF Streptococcus agar base Cat. No. 84633.0500
    Glass bottle
    30 mL
    1 * 30 mL
    1 * 30 mL
  • TTC 1% sterile solution
    TTC 1% sterile solution
    301950ZK
    Lactose TTC Tergitol® 7 agar Cat. No. 84657.0500 and Slanetz-Bartley agar (ISO) Cat. No. 84907.0500
    100 mL
    1 * 100 mL
    1 * 100 mL
  • Urea 40% solution
    Urea 40% solution
    992830ZF
    Urea agar base Cat. No. 84653.0500
    100 mL
    1 * 100 mL
    1 * 100 mL
  • Vancomycin selective supplement
    Vancomycin selective supplement
    928570NL