Japan
Order Entry
J.T.Baker® BAKER BIO-ANALYZED®, Buffer A: 20 mm NaOH in 10% Acetonitrile
J.T.Baker® BAKER BIO-ANALYZED®, Buffer A: 20 mm NaOH in 10% Acetonitrile
  9122-51
 :  Avantor
 :  
undefined
J.T.Baker® BAKER BIO-ANALYZED®, Buffer A: 20 mm NaOH in 10% Acetonitrile
  9122-51
 :  Avantor
 :  
 
 
   
  1 * 200 L

 

  • Concentration:
    20 mM NaOH
  • Pk:
    200 L
  • Description:
    Buffer A
  • Size:
    200 L