Japan
Order Entry
VWR®, Refractive index Certified Reference Material at 20, 25 and 30 °C
 
undefined
VWR®, Refractive index Certified Reference Material at 20, 25 and 30 °C
 

 

  •  
    Nominal refractive index at 25 °C
    Pack type
    Pk Info
    Pk
     
     
  • Refractive index certified reference material
    Refractive index certified reference material
    99177.100
    1.3325
    Glass bottle
    1×10 ml
    10 mL
       
       
    1 * 10 mL
    1 * 10 mL

  • Refractive index certified reference material
    Refractive index certified reference material
    99177.001
    1.3325
    Glass bottle
    5×10 ml
    1 SET
       
       
    1 * 1 SET
    1 * 1 SET

  • Refractive index certified reference material
    Refractive index certified reference material
    99178.100
    1.3891
    Glass bottle
    1×10 ml
    10 mL
       
       
    1 * 10 mL
    1 * 10 mL

  • Refractive index certified reference material
    Refractive index certified reference material
    99178.001
    1.3891
    Glass bottle
    5×10 ml
    1 SET
       
       
    1 * 1 SET
    1 * 1 SET

  • Refractive index certified reference material
    Refractive index certified reference material
    99179.100
    1.4023
    Glass bottle
    1×10 ml
    10 mL
       
       
    1 * 10 mL
    1 * 10 mL

  • Refractive index certified reference material
    Refractive index certified reference material
    99179.001
    1.4023
    Glass bottle
    5×10 ml
    1 SET
       
       
    1 * 1 SET
    1 * 1 SET

  • Refractive index certified reference material
    Refractive index certified reference material
    99180.100
    1.4196
    Glass bottle
    1×10 ml
    10 mL
       
       
    1 * 10 mL
    1 * 10 mL

  • Refractive index certified reference material
    Refractive index certified reference material
    99180.001
    1.4196
    Glass bottle
    5×10 ml
    1 SET
       
       
    1 * 1 SET
    1 * 1 SET

  • Refractive index certified reference material
    Refractive index certified reference material
    99181.100
    1.4206
    Glass bottle
    1×10 ml
    10 mL
       
       
    1 * 10 mL
    1 * 10 mL

  • Refractive index certified reference material
    Refractive index certified reference material
    99182.100
    1.4573
    Glass bottle
    1×10 ml
    10 mL
       
       
    1 * 10 mL
    1 * 10 mL

  • Refractive index certified reference material
    Refractive index certified reference material
    99183.100
    1.4941
    Glass bottle
    1×10 ml
    10 mL
       
       
    1 * 10 mL
    1 * 10 mL

  • Refractive index certified reference material
    Refractive index certified reference material
    99184.100
    1.5349
    Glass bottle
    1×10 ml
    10 mL
       
       
    1 * 10 mL
    1 * 10 mL

  • Refractive index certified reference material
    Refractive index certified reference material
    99185.100
    1.5440
    Glass bottle
    1×10 ml
    10 mL
       
       
    1 * 10 mL
    1 * 10 mL

  • Refractive index certified reference material
    Refractive index certified reference material
    99186.100
    1.6556
    Glass bottle
    1×10 ml
    10 mL
       
       
    1 * 10 mL
    1 * 10 mL