Masterflex® L/S® High-Performance Precision Pump Tubing, Tygon® A-60-F, Avantor®
Similar to Norprene® but with FDA approval.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Ideal for the food, dairy, and beverage industries
Characteristics: Heat, ozone, and UV light resistant, nonaging, nonoxidizing, superior acid and alkali resistance, opaque beige
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
- Environmentally Preferable:
- Model:
- 06402-36
- 06402-15
- 06402-35
- 06402-24
- Length per pack:
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- 15,2 m (50 ft.)
- Max. pressure:
- 1,4 bar (20 psi)
- 2,7 bar (40 psi)
- 2,4 bar (35 psi)
- 2,7 bar (40 psi)
- Min. temperature:
- –51 °C (–60 °F)
- –51 °C (–60 °F)
- –51 °C (–60 °F)
- –51 °C (–60 °F)
- Max. temperature:
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- 132 °C (270 °F)
- Min. temperature (dynamic):
- –20 °C (–4 °F)
- –20 °C (–4 °F)
- –20 °C (–4 °F)
- –20 °C (–4 °F)
- Max. temperature (dynamic):
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- 100 °C (212 °F)
- Max. vacuum:
- 610 mm Hg (24" Hg)
- 660 mm Hg (26" Hg)
- 660 mm Hg (26" Hg)
- 660 mm Hg (26" Hg)
- Pump series:
- L/S
- L/S
- L/S
- L/S
- Regulatory approvals:
- FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS
- FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS
- FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS
- FDA;Non-DEHP;Non-Phthalate;NSF-51;RoHS
- Sterilisation:
- Autoclave, Ethylene Oxide, Gamma Irradiation
- Autoclave, Ethylene Oxide, Gamma Irradiation
- Autoclave, Ethylene Oxide, Gamma Irradiation
- Autoclave, Ethylene Oxide, Gamma Irradiation
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- Yes
- Yes
- Yes
- Yes
- Sterilisation method gamma:
- Yes
- Yes
- Yes
- Yes
- Suction lift H₂O:
- 8,3 m (27 ft.)
- 8,8 m (29 ft.)
- 8,8 m (29 ft.)
- 8,8 m (29 ft.)
- Warranty:
- 1 year
- 1 year
- Tubing int.ØTubing materialL/S Masterflex tubing sizeHose barb sizeMaximum Flow RatePk
- L/S® High-Performance Precision Pump Tubing, Tygon® A-60-F, L/S 15; 50 ftL/S® High-Performance Precision Pump Tubing, Tygon® A-60-F, L/S 15; 50 ft4,80 mm (0,189")Tygon® A-60-F150,1875"1100 ml/min50 Feet1 * 50 Feet: MFLA06402-15CE certificate
Specifications
Model06402-15Environmentally PreferableLength per pack15,2 m (50 ft.)Max. pressure2,7 bar (40 psi)Min. temperature–51 °C (–60 °F)Max. temperature132 °C (270 °F)Min. temperature (dynamic)–20 °C (–4 °F)Max. temperature (dynamic)100 °C (212 °F)Max. vacuum660 mm Hg (26" Hg)Pump seriesL/SRegulatory approvalsFDA;Non-DEHP;Non-Phthalate;NSF-51;RoHSSterilisationAutoclave, Ethylene Oxide, Gamma IrradiationSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYesSuction lift H₂O8,8 m (29 ft.)Warranty1 year - L/S® High-Performance Precision Pump Tubing, Tygon® A-60-F, L/S 24; 50 ftL/S® High-Performance Precision Pump Tubing, Tygon® A-60-F, L/S 24; 50 ft6,40 mm (0,252")Tygon® A-60-F240,25"1800 ml/min1.524 cm1 * 50 Feet: MFLA06402-24CE certificate
Specifications
Model06402-24Environmentally PreferableLength per pack15,2 m (50 ft.)Max. pressure2,7 bar (40 psi)Min. temperature–51 °C (–60 °F)Max. temperature132 °C (270 °F)Min. temperature (dynamic)–20 °C (–4 °F)Max. temperature (dynamic)100 °C (212 °F)Max. vacuum660 mm Hg (26" Hg)Pump seriesL/SRegulatory approvalsFDA;Non-DEHP;Non-Phthalate;NSF-51;RoHSSterilisationAutoclave, Ethylene Oxide, Gamma IrradiationSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYesSuction lift H₂O8,8 m (29 ft.)Warranty1 year - L/S® High-Performance Precision Pump Tubing, Tygon® A-60-F, L/S 35; 50 ftL/S® High-Performance Precision Pump Tubing, Tygon® A-60-F, L/S 35; 50 ft7,90 mm (0,311")Tygon® A-60-F350,375"2600 ml/min50 Feet1 * 50 Feet: MFLA06402-35CE certificate
Specifications
Model06402-35Environmentally PreferableLength per pack15,2 m (50 ft.)Max. pressure2,4 bar (35 psi)Min. temperature–51 °C (–60 °F)Max. temperature132 °C (270 °F)Min. temperature (dynamic)–20 °C (–4 °F)Max. temperature (dynamic)100 °C (212 °F)Max. vacuum660 mm Hg (26" Hg)Pump seriesL/SRegulatory approvalsFDA;Non-DEHP;Non-Phthalate;NSF-51;RoHSSterilisationAutoclave, Ethylene Oxide, Gamma IrradiationSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYesSuction lift H₂O8,8 m (29 ft.)Warranty - L/S® High-Performance Precision Pump Tubing, Tygon® A-60-F, L/S 36; 50 ftL/S® High-Performance Precision Pump Tubing, Tygon® A-60-F, L/S 36; 50 ft9,70 mm (0,382")Tygon® A-60-F360,375"3400 ml/min50 Feet1 * 50 Feet: MFLA06402-36CE certificate
Specifications
Model06402-36Environmentally PreferableLength per pack15,2 m (50 ft.)Max. pressure1,4 bar (20 psi)Min. temperature–51 °C (–60 °F)Max. temperature132 °C (270 °F)Min. temperature (dynamic)–20 °C (–4 °F)Max. temperature (dynamic)100 °C (212 °F)Max. vacuum610 mm Hg (24" Hg)Pump seriesL/SRegulatory approvalsFDA;Non-DEHP;Non-Phthalate;NSF-51;RoHSSterilisationAutoclave, Ethylene Oxide, Gamma IrradiationSterilisation method autoclaveYesSterilisation method EtOYesSterilisation method gammaYesSuction lift H₂O8,3 m (27 ft.)Warranty