Pipette tips, VWR®
Supplier: VWR, part of Avantor
VWR®
VWRA612-6399EA 45.07 USD
VWRA612-6399 VWRA612-6264 VWRA612-6272 VWRA612-6196 VWRA612-6294 VWRA612-6274 VWRA612-6383 VWRA612-6195 VWRA612-6381 VWRA612-6432 VWRA612-6466 VWRA612-6191 VWRA612-6509 VWRA612-6464 VWRA612-6318 VWRA612-6190 VWRA612-6198 VWRA612-6503 VWRA612-6291 VWRA612-6488 VWRA612-6467 VWRA612-6534 VWRA612-6536 VWRA612-6284 VWRA612-6137 VWRA612-6249 VWRA612-6483 VWRA612-6148 VWRA612-6138 VWRA612-6144 VWRA612-6463 VWRA612-6496 VWRA612-6414 VWRA612-6405 VWRA612-6531 VWRA612-6332 VWRA612-6192 VWRA612-6598 VWRA612-6127 VWRA612-6333 VWRA612-6514 VWRA612-6474 VWRA612-6263 VWRA612-6523 VWRA612-6407 VWRA612-6384 VWRA612-6223 VWRA612-6491 VWRA612-6382 VWRA612-6484 VWRA612-6139 VWRA612-6247 VWRA612-6119 VWRA612-6529 VWRA612-6506 VWRA612-6182 VWRA612-6554 VWRA612-6179 VWRA612-6478 VWRA612-6126 VWRA612-6339 VWRA612-6189 VWRA612-6552 VWRA612-6489 VWRA612-6193 VWRA612-6520 VWRA612-6508 VWRA612-6292 VWRA612-6462 VWRA612-6473 VWRA612-6461 VWRA612-6221 VWRA612-6122 VWRA612-6521 VWRA612-6443 VWRA612-6197 VWRA612-6319 VWRA612-6419 VWRA612-6174 VWRA612-6493 VWRA612-6371 VWRA612-6434 VWRA612-6433 VWRA612-6408 VWRA612-6270 VWRA612-6261 VWRA612-6199 VWRA612-6380 VWRA612-6370 VWRA612-6177 VWRA612-6260 VWRA612-6460 VWRA612-6465 VWRA612-6372 VWRA612-6511 VWRA612-6660 VWRA612-6539 VWRA612-6481 VWRA612-6459 VWRA612-6354 VWRA612-6373 VWRA612-6128 VWRA612-6334 VWRA612-6124 VWRA612-6499 VWRA612-6262 VWRA612-6424 VWRA612-6504 VWRA612-6468 VWRA612-6403 VWRA612-6149 VWRA612-6293 VWRA612-6472 VWRA612-6580 VWRA612-6282 VWRA612-6159 VWRA612-6522 VWRA612-6406 VWRA612-6599 VWRA612-6470 VWRA612-6220 VWRA612-6458 VWRA612-6597 VWRA612-6246 VWRA612-6389 VWRA612-6213 VWRA612-6442 VWRA612-6486 VWRA612-6577 VWRA612-6513 VWRA612-6600 VWRA612-6129 VWRA612-6404 VWRA612-6469 VWRA612-6178 VWRA612-6143 VWRA612-6409 VWRA612-6348 VWRA612-6125 VWRA612-6388 VWRA612-6248 VWRA612-6471 VWRA612-6524 VWRA612-6544_U VWRA612-6118 VWRA612-6526 VWRA612-6476 VWRA612-6194 VWRA612-6142 VWRA612-6184 VWRA612-6212 VWRA612-6214 VWRA612-6579 VWRA612-6479 VWRA612-6123 VWRA612-6121 VWRA612-6553 VWRA612-6222 VWRA612-6494 VWRA612-6283 VWRA612-6173 VWRA612-6172
Pipette tips, VWR®
Pipette Tips
VWR® Pipettes tips are made from high-quality virgin polypropylene in a 100K cleanroom manufacturing facility.
  • Highly transparent tips, produced through high-quality diamond finish mold and special FDA approved medical grade virgin polypropylene
  • Innovative design for compatibility with a range of pipettes, ensuring minimum force requirement during loading and ejection
  • Ideal match for highly sensitive PCR works
  • Produced in a fully automatic, human touch-free, cleanroom production facility
  • Free from RNase, DNase, Human DNA, Pyrogen, and PCR inhibitor
  • Free of lubricants and heavy metals
  • Graduation marks on all tips act as visual checks to minimize the risk of errors during sample spiration
VWR® Pipette tips are available with wide ranges of attributes to meet specific needs of different applications. Bulk tips, racked tips, filter tips, low retention tips, individually wrapped tips, and reloadable tips are all available with universal compatibility.
Certifications: 100K cleanroom standard.
Order Now
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Learn more About VWR