ピペットチップ、VWR®製
供給業者: Avantor
VWR®
612-6127EA 623.84 USD
612-6127 612-6523 612-6248 612-6249 612-6128 612-6403 612-6524 612-6488 612-6521 612-6246 612-6125 612-6247 612-6126 612-6489 612-6522 612-6486 612-6123 612-6520 612-6124 612-6484 612-6121 612-6122 612-6529 612-6408 612-6409 612-6406 612-6407 612-6129 612-6404 612-6405 612-6526 612-6483 612-6481 612-6479 612-6513 612-6598 612-6478 612-6511 612-6599 612-6354 612-6597 612-6476 612-6198 612-6473 612-6474 612-6199 612-6514 612-6118 612-6119 612-6196 612-6471 612-6197 612-6472 612-6194 612-6195 612-6470 612-6192 612-6193 612-6190 612-6191 612-6468 612-6469 612-6348 612-6466 612-6467 612-6464 612-6189 612-6222 612-6223 612-6465 612-6462 612-6220 612-6463 612-6221 612-6509 612-6508 612-6506 612-6503 612-6504 612-6460 612-6461 612-6580 612-6184 612-6182 612-6458 612-6579 612-6334 612-6213 612-6577 612-6214 612-6332 612-6178 612-6333 612-6179 612-6212 612-6177 612-6459 612-6339 612-6174 612-6293 612-6172 612-6294 612-6173 612-6291 612-6292 612-6600 612-6442 612-6443 612-6284 612-6282 612-6283 612-6554 612-6433 612-6434 612-6159 612-6552 612-6399 612-6432 612-6553 612-6318 612-6319 612-6274 612-6272 612-6270 612-6424 612-6149 612-6389 612-6148 612-6388 612-6264 612-6660 612-6143 612-6144 612-6383 612-6262 612-6263 612-6384 612-6142 612-6381 612-6260 612-6382 612-6261 612-6380 612-6534 612-6138 612-6414 612-6139 612-6499 612-6137 612-6531 612-6496 612-6419 612-6539 612-6536 612-6493 612-6372 612-6373 612-6494 612-6491 612-6370 612-6371
ピペットチップ、VWR®製
ピペットチップ
VWR® Pipettes tips are made from high-quality virgin polypropylene in a 100K cleanroom manufacturing facility.
  • Highly transparent tips, produced through high-quality diamond finish mold and special FDA approved medical grade virgin polypropylene
  • Innovative design for compatibility with a range of pipettes, ensuring minimum force requirement during loading and ejection
  • Ideal match for highly sensitive PCR works
  • Produced in a fully automatic, human touch-free, cleanroom production facility
  • Free from RNase, DNase, Human DNA, Pyrogen, and PCR inhibitor
  • Free of lubricants and heavy metals
  • Graduation marks on all tips act as visual checks to minimize the risk of errors during sample spiration
VWR® Pipette tips are available with wide ranges of attributes to meet specific needs of different applications. Bulk tips, racked tips, filter tips, low retention tips, individually wrapped tips, and reloadable tips are all available with universal compatibility.
Through strict adherence to ISO 9001 compliant manufacturing standards, VWR® pipette tips ensure the elimination of external contamination sources, resulting in unmatched quality. Rely on the excellence of VWR® pipette tips to elevate performance and repeatability, surpassing all competing alternatives.
証明:100K cleanroom standard.
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR