Japan
Order Entry
VWR®, Cryobox, Light-Sensitive
VWR®, Cryobox, Light-Sensitive
Catalog # VWRU80080-597
Supplier:  VWR Collection
undefined
VWR®, Cryobox, Light-Sensitive
Catalog # VWRU80080-597
Supplier:  VWR Collection
Price
Inquire For Price
Log in for availability
Unit of Measure 1 * 1 items

Specifications

  • Compartments:
    10×10
  • For:
    Microtubes up to 2,0 ml
  • W×D×H ext.:
    140×140×60 mm
  • Colour:
    Black
  • Pk:
    1

Specifications

About this item

PP, black.

  • The reinforced seal, triple hinges and locking claps keeps samples secure
  • Alpha-numeric grid on the exterior and interior of the box
  • Stacking rings and a matte surface suitable for printing and labelling

Ideal for light-sensitive specimens and suitable for micro-tubes and other cryogenic vials.