Racks for microcentrifuge tubes, 96-place, VWR®
供給業者: VWR Collection
VWRU30128-346EA 59.92 USD
VWRU30128-346
Racks for microcentrifuge tubes, 96-place, VWR®
PP, double sided.
  • Use with 0,5/1,7 ml/2 ml microtubes and 2 ml cryotubes
  • Hold 1,7 ml and 2 ml tubes on one side and 0,5 ml on the reverse side
  • Suitable for freezer use down to −80 °C
  • Autoclavable
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR