VWR® Premium Line, Serological Pipettes
Supplier: VWR Collection
Certificates
About this item
PS, transparent, graduated, non sterile or sterile. These disposable pipettes for liquid transfers are made of GPPS (general polystyrene), in compliance with USP Class VI.
- DNase-/RNase-free, non pyrogenic, non cytotoxic, non haemolytic
- Meet the requirement of BST/TSE; latex-free
- Graduations are calibrated for accurate dispensing to within ±2%
- Pipettes are colour coded according to their volume
- Sterile pipettes: Sterilised by irradiation
- Printed with a batch number for traceability
Manufactured in a class 100 000 cleanroom environment. Manufactured under EN ISO 13485:2003 and ISO 9001:2008 quality management systems.
Packaging: Sterile pipettes are available either individually packed in peel-to-open wrap or plastic-plastic wrap or plastic packed in vacuum PE bag with zip lock (10/25/50 pipettes/bag). Non sterile pipettes are vacuum bulk packed in PE bags.
Specifications
- Catalog No:
- 612-5828
- 612-5827
- 612-5826
- 612-5543
- 612-5833
- 612-5539
- 612-5540
- 612-5541
- 612-5542
- 612-5507
- 612-5521
- 612-5522
- 612-5523
- 612-5506
- 612-5832
- 612-5830
- 612-5836
- 612-5834
- 612-5829
- 612-5835
- 612-5831
- 612-5837
- 612-5546
- 612-5547
- 612-5548
- 612-5504
- 612-5544
- 612-5505
- 612-5545
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
- ISO 14001/Certificate No.: 04923E00243R0M
Specifications
Related Information
Certificates
Search by Lot NumberAvantor Cell Culture Solutions.pdf VWR® for Liquid Handling VWR® Serological Pipettes
Product Literature
CoC_VWRI612-5504_180923067 CoC_VWRI612-5504_181112067 CoC_VWRI612-5504_190422067 CoC_VWRI612-5504_191215067 CoC_VWRI612-5504_200423067
More
TDS VWR® Serological Pipettes
Datasheet
Sterile, individually packed in peel-to-open wraps
Product Information
- Catalog #CapacityDivisionSterileLengthColour codePackedAvailabilityPrice
- StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Short formSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Short formSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Short formSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StandardSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StandardSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StandardSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact
Sterile, individually wraped, plastic/plastic
Product Information
- Catalog #CapacityDivisionSterileLengthColour codePackedAvailabilityPrice
- StandardSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Short formSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StandardSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StandardSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact
Sterile, 10/25 or 50 pipettes per PE bag with zip lock
Product Information
- Catalog #CapacityDivisionSterileLengthColour codePackedAvailabilityPrice
- Short formSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Wide tipSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StandardSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StandardSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StandardSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact
Non sterile, bulk packed in PE bags
Product Information
- Catalog #CapacityDivisionSterileLengthColour codePackedAvailabilityPrice
- StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - Short formSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact - StretchedSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactEnvironmental certificationISO 14001/Certificate No.: 04923E00243R0MLow Manufacturing Impact