Automated pipetting systems, epMotion® 5070
供給業者: Eppendorf
epMotion®
EPPE5070000.719 EPPE5070000.140 EPPE5070000.700 631-0193
Automated pipetting systems, epMotion® 5070
Automated Liquid Handling Systems
The Eppendorf epMotion® 5070 automated pipetting systems are high-performance workstations that are capable of handling all pipetting and dispensing tasks in the laboratory.
  • This automated pipetting system includes a four position deck for standard 96/384 well microplates
  • Convenient control with the remote control unit
  • Easy programming with the compact control panel and keyboard, mouse and colour monitor
  • Optical sensor for identifying laboratory equipment, tips and reagents
  • Dispensing system based on air displacement ceramic pistons requires no maintenance
  • Extremely precise, interchangeable pipetting heads, single-channel or eight-channel, autoclavable at 121 °C, offering three volume ranges (1 to 50 µl, 20 to 300 µl, 40 to 1000 µl)
The Eppendorf epMotion® 5070 is the most compact version, for precise, reproducible pipetting. It is ideal for routine applications such as series dilutions, dispensing reagents, transferring samples from tubes to plates, and standardising samples. It is equipped with a safety hood with a motorised front door and is compatible with PCR microplates and plates with up to 384 wells and test tube sizes from 0,2 to 50 ml.
The volumetric dispensing systems can be integrated easily in the work environment and provide an economical solution for these time-consuming, delicate jobs, so that personnel can spend more time on other laboratory work. The modular design of the entire epMotion® range also lends itself to a wide variety of options, extending its implementation possibilities enormously.
注文情報:注文情報:注文情報:Eppendorf epMotion® 5070 automated pipetting systems are supplied with a control unit, MotionManager v2,0 or higher software, optical sensor, one waste container, MultiMediaCard (MMC) with reader, and user's manual. Pipetting heads, pipette tips and reagent containers may be purchased separately. Other accessories are available on request.
今すぐ注文
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
詳細確認 About VWR