Order Entry
Accessories for VWR® SAS super Isolator
undefined
Accessories for VWR® SAS super Isolator
  •  
    Pk
  • Aluminium adapter for Petri dishes
    Aluminium adapter for Petri dishes
    710-0974
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Aspirating chamber for contact plates, Ø 55 mm
    Aspirating chamber for contact plates, Ø 55 mm
    710-0947
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Aspirating chamber for Petri dishes, Ø 90 mm
    Aspirating chamber for Petri dishes, Ø 90 mm
    710-0948
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Command unit for aspirating chamber
    Command unit for aspirating chamber
    PBIB91019
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Extension cable (3 m)
    Extension cable (3 m)
    710-0968
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Fixed wall-type extension cable
    Fixed wall-type extension cable
    710-1983
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • In-Out MH spare battery
    In-Out MH spare battery
    710-0998
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Interface cable
    Interface cable
    710-0964
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • IQ/OQ/PQ manual
    IQ/OQ/PQ manual
    710-0954
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Plug coupler
    Plug coupler
    710-1980
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Remote control
    Remote control
    710-0962
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Removable wall-type extension cable
    Removable wall-type extension cable
    710-1984
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Software
    Software
    710-0975
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable