Order Entry
VWR Chemicals, Base Materials for Dehydrated Media
VWR Chemicals, Base Materials for Dehydrated Media
Supplier:  VWR Chemicals
Certificates
undefined
VWR Chemicals, Base Materials for Dehydrated Media
Supplier:  VWR Chemicals
Certificates
Product Information
  • Catalog #
    Pack type
    Pk
    Availability
    Price
  • Bacteriological agar
    Bacteriological agar
    84609.5000
    5 kg
    Log in for availability
    1 * 5 kg
    1 * 5 kg
  • Bacteriological agar
    Bacteriological agar
    84609.0500
    Plastic bottle
    500 g
    Log in for availability
    1 * 500 g
    1 * 500 g
  • Casein hydrolysate (acid)
    Casein hydrolysate (acid)
    84623.0500
    500 g
    Log in for availability
    1 * 500 g
    1 * 500 g
  • Casein hydrolisate acid
    Casein hydrolisate acid
    85862.0500
    500 g
    Log in for availability
    1 * 500 g
    1 * 500 g
  • Casein hydrolisate acid
    Casein hydrolisate acid
    85862.5000
    5 kg
    Log in for availability
    1 * 5 kg
    1 * 5 kg
  • Malt extract
    Malt extract
    84618.0500
    500 g
    Log in for availability
    1 * 500 g
    1 * 500 g
  • Peptone, meat (enzymatic digest of animal tissue)
    Peptone, meat (enzymatic digest of animal tissue)
    84620.0500
    500 g
    Log in for availability
    1 * 500 g
    1 * 500 g
  • Meat extract dry
    Meat extract dry
    84688.0500
    500 g
    Log in for availability
    1 * 500 g
    1 * 500 g
  • Meat extract dry
    Meat extract dry
    84688.5000
    5 kg
    Log in for availability
    1 * 5 kg
    1 * 5 kg
  • Hiveg pea hydrolysat
    Hiveg pea hydrolysat
    84862.25000
    25 kg
    Log in for availability
    1 * 25 kg
    1 * 25 kg
  • Peptone, meat (enzymatic digest of animal tissue)
    Peptone, meat (enzymatic digest of animal tissue)
    84620.5000
    5 kg
    Log in for availability
    1 * 5 kg
    1 * 5 kg
  • Peptone, meat
    Peptone, meat
    84687.0500
    500 g
    Log in for availability
    1 * 500 g
    1 * 500 g
  • Skimmed milk powder
    Skimmed milk powder
    84615.0500
    500 g
    Log in for availability
    1 * 500 g
    1 * 500 g
  • Yeast extract
    Yeast extract
    5 kg
    Log in for availability
    1 * 5 kg
    1 * 5 kg
  • Soya peptone (enzymatic digest of soybean meal)
    Soya peptone (enzymatic digest of soybean meal)
    84616.0500
    500 g
    Log in for availability
    1 * 500 g
    1 * 500 g
  • Tryptone (peptone from casein)
    Tryptone (peptone from casein)
    84610.0500
    500 g
    Log in for availability
    1 * 500 g
    1 * 500 g
  • Tryptone (peptone from casein)
    Tryptone (peptone from casein)
    84610.5000
    5 kg
    Log in for availability
    1 * 5 kg
    1 * 5 kg
  • Yeast extract
    Yeast extract
    84601.0500
    500 g
    Log in for availability
    1 * 500 g
    1 * 500 g
  • Yeast extract
    Yeast extract
    84601.5000
    5 kg
    Log in for availability
    1 * 5 kg
    1 * 5 kg
  • Peptone (pancreatic digest), indol-free for bacteriology
    Peptone (pancreatic digest), indol-free for bacteriology
    1 kg
    Log in for availability
    1 * 1 kg
    1 * 1 kg
  • Skimmed milk powder
    Skimmed milk powder
    84615.5000
    5 kg
    Log in for availability
    1 * 5 kg
    1 * 5 kg