Order Entry
VWR® Q Path® Filing Cabinet for Cassettes
VWR® Q Path® Filing Cabinet for Cassettes
  631-1065
undefined
VWR® Q Path® Filing Cabinet for Cassettes
  631-1065
  1 * 2 items
  • Colour:
    White
  • Description:
    Filing cabinets
  • W×D×H:
    315×255×120 mm
  • Pk:
    2
Filing cabinet in white cardboard.
  • Supplied assembled
  • Capacity for 410 cassettes
Two drawers.