Avantor® ACE® SuperC18, HPLC Columns, 5 µm
Supplier: Avantor
Certificates
About this item
Avantor® ACE® SuperC18 columns provide high stability across an extended pH range. These stainless steel columns are ideal for use with LC/MS compatible buffers and can be used with both MeOH and MeCN mobile phases.
- Ultra-inert for maximum performance and reproducibility
- Ultra low bleed for improved LC/MS compatibility
- Wide range of particle sizes (1,7 to 10 µm) and complementary phases
Maximum recommended pH: 1,5 to 11,5
Max. pressure: 275 bar
Specifications
- Catalog No:
- ACE-1211-15005
- ACE-1211-3030
- ACE-1211-0510
- ACE-1211-0520
- ACE-1211-2530
- ACE12110110GD
- ACE-1211-1530
- ACE12110120GD
- ACE-1211-1030
- ACE-1211-1520
- ACE-1211-2510
- ACE-1211-2520
- ACE-1211-0102GD
- ACE-1211-1010
- ACE-1211-1510
- ACE-1211-7530
- ACE-1211-1020
- ACE-1211-0103GD
- ACE-1211-0530
- ACE-1211-1220
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- 14,8%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Max. pressure:
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- 275 bar
- pH range:
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- 1,5 - 11,5
- Surface area:
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- 400 m²/g
- USP code:
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- L1
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
Specifications
Product Information
- Catalog #Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
- 1/16" connections1/16" connectionsACE-1211-15005MicroboreSuperC18™90 Å5 µm0,50 mm150 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Environmentally PreferableCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1211-1010Semi-preparativeSuperC18™90 Å5 µm10,00 mm100 mmLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 1/16" connections1/16" connectionsACE-1211-1510Semi-preparativeSuperC18™90 Å5 µm10,00 mm150 mmLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 1/16" connections1/16" connectionsACE-1211-2510Semi-preparativeSuperC18™90 Å5 µm10,00 mm250 mmLog in for availability1 * 1 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 1/16" connections1/16" connectionsACE-1211-0520PreparativeSuperC18™90 Å5 µm21,20 mm50 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Environmentally PreferableCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1211-1020PreparativeSuperC18™90 Å5 µm21,20 mm100 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Environmentally PreferableCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1211-1520PreparativeSuperC18™90 Å5 µm21,20 mm150 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Environmentally PreferableCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1211-2520PreparativeSuperC18™90 Å5 µm21,20 mm250 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Environmentally PreferableCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1211-2530PreparativeSuperC18™90 Å5 µm30,00 mm250 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Environmentally PreferableCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1211-3030PreparativeSuperC18™90 Å5 µm30,00 mm300 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Environmentally PreferableCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certification - Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon load14,8%EndcappedYesMax. pressurepH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - 1/16" connections1/16" connectionsACE-1211-1530PreparativeSuperC18™90 Å5 µm30,00 mm150 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Environmentally PreferableCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1211-1030PreparativeSuperC18™90 Å5 µm30,00 mm100 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Environmentally PreferableCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1211-7530PreparativeSuperC18™90 Å5 µm30,00 mm75 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Environmentally PreferableCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certification - 1/16" connections1/16" connectionsACE-1211-0530PreparativeSuperC18™90 Å5 µm30,00 mm50 mmLog in for availability1 * 1 itemsSpecifications:
More
Specifications
Environmentally PreferableCarbon load14,8%EndcappedYesMax. pressure275 barpH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certification - Specifications:
More
Specifications
Environmentally PreferableCarbon load14,8%EndcappedYesMax. pressurepH range1,5 - 11,5Surface area400 m²/gUSP codeL1Environmental certification
Guard cartridges
Product Information
- Catalog #Column FormatPhasePore sizeParticle SizeØ int.LengthAvailabilityPrice
- For preparative columnsFor preparative columnsACE12110110GDGuard cartridgeSuperC18™90 Å5 µmLog in for availability1 * 3 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedMax. pressurepH rangeSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - Specifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedMax. pressurepH rangeSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - For analytical columns (I.Ø 2,1 mm)For analytical columns (I.Ø 2,1 mm)ACE-1211-0102GDGuard cartridgeSuperC18™90 Å5 µmLog in for availability1 * 5 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedMax. pressurepH rangeSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - For analytical columns (I.Ø 3,0, 4,0, 4,6 mm)For analytical columns (I.Ø 3,0, 4,0, 4,6 mm)ACE-1211-0103GDGuard cartridgeSuperC18™90 Å5 µmLog in for availability1 * 5 itemsSpecifications:
Low Manufacturing Impact More
Specifications
Environmentally PreferableLow Manufacturing ImpactCarbon loadEndcappedMax. pressurepH rangeSurface areaUSP codeEnvironmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact