VWR®, Filter Funnels according to Dr. Hirsch
Supplier: VWR Collection
Certificates
About this item
Porcelain, glased inside and out, except rim.
Funnel with integral perforated filter plate can be used with paper or membrane discs.
Product Information
- Catalog #Supplier #LengthFor filter ØFunnel O-ØPkAvailabilityPrice
- Specifications:
Specifications
Environmentally Preferable - Specifications:
Specifications
Environmentally Preferable - Specifications:
Specifications
Environmentally Preferable