VWR® Talon®, S-connectors
S-connectors made from electro-polished stainless steel or nickel plated zinc.
- Clamp is ideal for constructing lab frames or other supports requiring the connection of two perpendicular rods
- Clamp connects two 13 mm rods at a 90° angle and features separate adjustment screws for each rod location
- MaterialFor rod ØPk
- S-connector
Specifications
Environmentally Preferable - S-connector
Specifications
Environmentally Preferable