Digital Solutions
Applications
Support
About
Order Entry
VWR®, Screw Caps for Microcentrifuge Tubes
:
VWR Collection
VWR®, Screw Caps for Microcentrifuge Tubes
:
VWR Collection
Colour
Pk
Standard screw cap
Standard screw cap
VWRU89004-354
89004-354
Red
500
1 * 500 items
-
+
1 * 500 items
-
+
Specifications
Environmentally Preferable
Standard screw cap
Standard screw cap
VWRU89004-362
89004-362
Assorted
500
1 * 500 items
-
+
1 * 500 items
-
+
Specifications
Environmentally Preferable