Digital Solutions
Applications
Support
About
Order Entry
VWR® Enviro Safe Dry Block/Incubator Thermometers
:
VWR Collection
VWR® Enviro Safe Dry Block/Incubator Thermometers
:
VWR Collection
AT-VWR Thermometer FAQs
VWR® for Temperature Measurement and Recording
Range
Accuracy
Division
Length
Filling
Immersion depth
Pk
620-0790
+24…+57 °C
±0,5 °C
0,5 °C
175 mm
Enviro safe
35 mm
1
1 * 1 items
-
+
1 * 1 items
-
+
: VWRI620-0790
Specifications
Environmentally Preferable
620-0791
+24…+57 °C
±0,5 °C
0,5 °C
175 mm
Enviro safe
35 mm
1
1 * 1 items
-
+
1 * 1 items
-
+
: VWRI620-0791
Specifications
Environmentally Preferable