Fully encapsulated suits with positive pressure provided by powered air purifying respirators (PAPR). Made from a microporous PP laminate with a non woven inner layer.
- Double-elasticated cuffs: Enables a liquid-tight connection with chemical protective glove
- Air permeable SMS collar maintains sufficient air in the breathing zone whilst allowing excess air to flow into the body of the suit
- Attached socks with elasticated boot over-flap
- Panoramic visor design ensures a good field of vision for the wearer
- Emergency rip cord permits rapid doffing of the suit in cases of emergency or undue distress to the wearer
- Four exhalation valves fitted to the rear of the suit allow CO₂ to escape and equalise pressure within the suit
- Stitched and taped seams increase strength and make an effective liquid and particle barrier
For use with Scott Proflow SC/EX fan units and filters.
Ideal for use in hazardous environments where a high level of liquid chemical and/or particle protection is required combined with a need for comfortable working conditions for the wearer.
Typical applications include: High risk areas, handling Active Pharmaceutical Ingredients (APIs) and potential sensitising agents.
証明:Type 3-B/4-B/5-B, EN 1073-2, EN 1149-1, EN 14126, EN 12941.
注文情報:注文情報:注文情報:Scott fan units, breathing hoses and filters are not included.