Order Entry
J.T.Baker® BAKERBOND®, Wide-Pore Prep Packings
J.T.Baker® BAKERBOND®, Wide-Pore Prep Packings
Supplier:  Avantor
Certificates
undefined
J.T.Baker® BAKERBOND®, Wide-Pore Prep Packings
Supplier:  Avantor
Certificates
Specifications
  • Working pH:
    2 - 8,5
Specifications
Related Information
Ion exchange
Product Information
  • Catalog #
    Particle Size
    Pore size
    Pk
    Availability
    Price
  • PEI and quaternary amine
    PEI and quaternary amine
    7183-02
    15 µm
    300 Å
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g
Reverse phase
Product Information
  • Catalog #
    Particle Size
    Pore size
    Pk
    Availability
    Price
  • Octadecyl (C₁₈), BAKERBOND®, PE bottle
    Octadecyl (C₁₈), BAKERBOND®, PE bottle
    15 µm
    300 Å
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g
  • Octadecyl (C₁₈), BAKERBOND®, PE bottle
    Octadecyl (C₁₈), BAKERBOND®, PE bottle
    15 µm
    300 Å
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g
  • Octadecyl (C₁₈), BAKERBOND®, PE bottle
    Octadecyl (C₁₈), BAKERBOND®, PE bottle
    7207-00
    15 - 30 µm
    300 Å
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g