Order Entry
J.T.Baker® BAKERBOND®, Reversed Phase Bulk Packing Material for Preperative HPLC Columns, WP Butyl C4
J.T.Baker® BAKERBOND®, Reversed Phase Bulk Packing Material for Preperative HPLC Columns, WP Butyl C4
Supplier:  Avantor
Certificates
undefined
J.T.Baker® BAKERBOND®, Reversed Phase Bulk Packing Material for Preperative HPLC Columns, WP Butyl C4
Supplier:  Avantor
Certificates
Specifications
  • Average particle size:
    15 µm
  • Working pH:
    2 - 8,5
Specifications
Related Information
Product Information
  • Catalog #
    Particle Size
    Pore size
    Pk
    Availability
    Price
  • Silica-based reverse phase media
    Silica-based reverse phase media
    7179-00
    15 µm
    300 Å
    100 g
    Log in for availability
    Log in for availability
    4 * 100 g/Case
    4 * 100 g/Case
  • Silica-based reverse phase media
    Silica-based reverse phase media
    15 µm
    300 Å
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g
  • Silica-based reverse phase media
    Silica-based reverse phase media
    15 µm
    300 Å
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g