Order Entry
VWR® Buffer Tablets for Haematology
VWR® Buffer Tablets for Haematology
undefined
VWR® Buffer Tablets for Haematology
Supplier:  VWR Chemicals
Certificates
  •  
    pH
    Final volume
    Pk
  • Buffer tablets
    Buffer tablets
    331932D
    6,8
    100 ml
    50 Tab.
    1 * 50 Tablet
    1 * 50 Tablet
  • Buffer tablets
    Buffer tablets
    331552S
    7,0 ±0,02
    100 ml
    50 Tab.
    1 * 50 Tablet
    1 * 50 Tablet
  • Buffer tablets
    Buffer tablets
    331942F
    7,2
    100 ml
    50 Tab.
    1 * 50 Tablet
    1 * 50 Tablet
  • Buffer tablets
    Buffer tablets
    331562U
    9,2 ±0,02
    100 ml
    50 Tab.
    1 * 50 Tablet
    1 * 50 Tablet