Avantor® Genesis, HPLC Columns, C8
: HICHROM
Avantor® Genesis phases are based on high purity metal-free spherical silica. They are suitable for the analysis of a wide range of analytes.
- Excellent peak shape and efficiency
- Long column life
- Catalog No:
- FM25962E
- FM25964E
- GEN-5C8-150D
- FM15962E
- FM5964
- FK5962E
- FH1962-2/N
- FM10962E
- HICHGEN-4C8-125DF
- FK25964E
- GEN-5C8-250D
- Description:
- C8
- C8 EC
- C8
- C8
- C8 EC
- C8
- C8
- C8
- C8
- C8 EC
- C8
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Carbon load:
- 11%
- 11%
- 11%
- 11%
- 11%
- 11%
- 11%
- 11%
- 11%
- 11%
- 11%
- Endcapped:
- No
- Yes
- No
- No
- Yes
- No
- No
- No
- No
- Yes
- No
- pH range:
- 1 - 10
- 1 - 10
- 1 - 10
- 1 - 10
- 1 - 10
- 1 - 10
- 1 - 10
- 1 - 10
- 1 - 10
- 1 - 10
- 1 - 10
- Surface area:
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- 300 m²/g
- USP code:
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- L7
- Environmental certification:
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- ISO 14001:2015 (530840 UM15)
- Column FormatPhaseColumn hardwarePore sizeParticle SizeØ int.Length
- : HICHFK5962E
Specifications
DescriptionC8Environmentally PreferableCarbon load11%EndcappedNopH range1 - 10Surface area300 m²/gUSP codeL7Environmental certification - : HICHGEN-4C8-125DF
Specifications
DescriptionC8Environmentally PreferableCarbon load11%EndcappedNopH range1 - 10Surface area300 m²/gUSP codeL7Environmental certification - : HICHFM10962E
Specifications
DescriptionC8Environmentally PreferableCarbon load11%EndcappedNopH range1 - 10Surface area300 m²/gUSP codeL7Environmental certification - : HICHFM15962E
Specifications
DescriptionC8Environmentally PreferableCarbon load11%EndcappedNopH range1 - 10Surface area300 m²/gUSP codeL7Environmental certification - : HICHGEN-5C8-150D
Specifications
DescriptionC8Environmentally PreferableCarbon load11%EndcappedNopH range1 - 10Surface area300 m²/gUSP codeL7Environmental certification - : HICHGEN-5C8-250D
Specifications
DescriptionC8Environmentally PreferableCarbon load11%EndcappedNopH range1 - 10Surface area300 m²/gUSP codeL7Environmental certification - : HICHFM25962E
Specifications
DescriptionC8Environmentally PreferableCarbon load11%EndcappedNopH range1 - 10Surface area300 m²/gUSP codeL7Environmental certification - : HICHFK25964E
Low Manufacturing Impact Specifications
DescriptionC8 ECEnvironmentally PreferableLow Manufacturing ImpactCarbon load11%EndcappedYespH range1 - 10Surface area300 m²/gUSP codeL7Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - : HICHFM5964
Low Manufacturing Impact Specifications
DescriptionC8 ECEnvironmentally PreferableLow Manufacturing ImpactCarbon load11%EndcappedYespH range1 - 10Surface area300 m²/gUSP codeL7Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact - : HICHFM25964E
Low Manufacturing Impact Specifications
DescriptionC8 ECEnvironmentally PreferableLow Manufacturing ImpactCarbon load11%EndcappedYespH range1 - 10Surface area300 m²/gUSP codeL7Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact
Guard cartridges
- Column FormatPhaseColumn hardwarePore sizeParticle SizeØ int.Length
- : HICHFH1962-2/N
Low Manufacturing Impact Specifications
DescriptionC8Environmentally PreferableLow Manufacturing ImpactCarbon load11%EndcappedNopH range1 - 10Surface area300 m²/gUSP codeL7Environmental certificationISO 14001:2015 (530840 UM15)Low Manufacturing Impact