Order Entry
VWR®, Cryoboxes, Half Size, 136×66,5 mm
undefined
VWR®, Cryoboxes, Half Size, 136×66,5 mm
Supplier:  VWR Collection
Cryoboxes, half size, with dividers
Product Information
  • Catalog #
    Compartments
    Colour
    W×D×H ext.
    Pk
    Availability
    Price
  • 479-7000
    5×10
    White
    136×66,5×50 mm
    10
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • 479-7001
    5×10
    Blue
    136×66,5×50 mm
    10
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • 479-7002
    5×10
    Red
    136×66,5×50 mm
    10
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • 479-7003
    5×10
    Green
    136×66,5×50 mm
    10
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
  • 479-7004
    5×10
    Yellow
    136×66,5×50 mm
    10
    Log in for availability
    1 * 10 items
    1 * 10 items
    Specifications:
    Accessories
    Specifications
    Environmentally Preferable
Dividers for cryoboxes half size
  •  
    Seq
    Compartments
    Colour
    W×D×H ext.
    Pk
  • 06
    5×10
    White
    136×66,5×50 mm