Digital Solutions
Applications
Support
About
Order Entry
VWR®, Dual-Purpose Scoops, Sterile and Non Sterile
:
VWR Collection
VWR®, Dual-Purpose Scoops, Sterile and Non Sterile
:
VWR Collection
Avantor Cell Culture Solutions.pdf
VWR® Collection Selection Catalogue
Type
Length
Width
Sterile
Colour
Pk
611-4350
Small
127 mm
63 mm
-
White
100
1 * 100 items
-
+
1 * 100 items
-
+
: VWRI611-4350
Specifications
Environmentally Preferable
611-4349
Large
127 mm
90 mm
-
White
100
1 * 100 items
-
+
1 * 100 items
-
+
: VWRI611-4349
Specifications
Environmentally Preferable