VWR®, Sucrose (Brix) standards, stabilised solutions for refractometry
A range of Brix/refractive index standards for ease of use when calibrating all types of refractometers.
- Manufactured using high purity raw materials
- Contain the same raw materials as the ICUMSA range, but are stabilised to have an extended shelf life of 12 months
These products represent excellent value for users that are not required to follow ICUMSA guidelines.
Note: The Brix scale is shown with the degree type symbol (x°) but it equates to the % concentration of the stabilised sucrose solution used.
Therefore 0° = 0% concentration
5° = 5% concentration
* The refractive index value in the table below refers to the nominal refractive index, and is taken from the ICUMSA published tables for all product numbers except 87022.015 (Brix value 67,5°) which is a measured value.
Product Information
- Catalog #Refractive indexAccuracyPkAvailabilityPrice
- Sucrose (Brix) standard, stabilised, 0°Sucrose (Brix) standard, stabilised, 0°87000.0151,332986± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 5°Sucrose (Brix) standard, stabilised, 5°87001.0151,340264± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 7°Sucrose (Brix) standard, stabilised, 7°87002.0151,343253± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 10°Sucrose (Brix) standard, stabilised, 10°87003.0151,347824± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 11,2°Sucrose (Brix) standard, stabilised, 11,2°87004.0151,349682± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 12°Sucrose (Brix) standard, stabilised, 12°87006.0151,350930± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 12,5°Sucrose (Brix) standard, stabilised, 12,5°87007.0151,351714± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 14,9°Sucrose (Brix) standard, stabilised, 14,9°87008.0151,355519± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 15°Sucrose (Brix) standard, stabilised, 15°87009.0151,355679± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 20°Sucrose (Brix) standard, stabilised, 20°87011.0151,363842± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 25°Sucrose (Brix) standard, stabilised, 25°87012.0151,372328± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 30°Sucrose (Brix) standard, stabilised, 30°87013.0151,381149± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 35°Sucrose (Brix) standard, stabilised, 35°87014.0151,390322± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 40°Sucrose (Brix) standard, stabilised, 40°87015.0151,399860± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 45°Sucrose (Brix) standard, stabilised, 45°87016.0151,409777± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 50°Sucrose (Brix) standard, stabilised, 50°87018.0151,420087± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 55°Sucrose (Brix) standard, stabilised, 55°87019.0151,430800± 0,11°15 mL1 * 15 mL
- Sucrose (Brix) standard, stabilised, 60°Sucrose (Brix) standard, stabilised, 60°87021.0151,441928± 0,11°15 mL1 * 15 mL