Order Entry
VWR®, Flat Screw Caps for Microcentrifuge Tubes
undefined
VWR®, Flat Screw Caps for Microcentrifuge Tubes
  • Catalog No:
  • 525-1179
  • 525-1181
  • 525-1180
  • 525-1183
  • 525-1182
  • 525-1185
  • 525-1184
  • 525-1187
  • 525-1186
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
  •  
    Colour
    Pk
  • 525-1187
    Assorted
    500
    1 * 500 items
    1 * 500 items
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 525-1180
    Blue
    500
    1 * 500 items
    1 * 500 items
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 525-1179
    Green
    500
    1 * 500 items
    1 * 500 items
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 525-1186
    Natural
    500
    1 * 500 items
    1 * 500 items
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 525-1183
    Orange
    500
    1 * 500 items
    1 * 500 items
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 525-1181
    Purple
    500
    1 * 500 items
    1 * 500 items
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 525-1182
    Red
    500
    1 * 500 items
    1 * 500 items
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 525-1185
    White
    500
    1 * 500 items
    1 * 500 items
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • 525-1184
    Yellow
    500
    1 * 500 items
    1 * 500 items
      Low Manufacturing Impact
    Specifications
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact