Order Entry
Rankem®, Erlenmeyer Flasks, Narrow Neck, Glass
 :  Transene
Rankem®, Erlenmeyer Flasks, Narrow Neck, Glass
 :  Transene
  •  
    Capacity
    Height
    Socket size
    Pk
  • RANKFE-NMIC-0025
    FE-NMIC-0025
    25 ml
    70 mm
    14/23 NS
    12
    12 * 12 items/Case
    12 * 12 items/Case
    Specifications
    Environmentally Preferable
  • RANKFE-NMIC-0100
    FE-NMIC-0100
    100 ml
    100 mm
    24/29 NS
    12
    12 * 12 items/Case
    12 * 12 items/Case
    Specifications
    Environmentally Preferable
  • RANKFE-NMIC-0250
    FE-NMIC-0250
    250 ml
    140 mm
    24/29 NS
    12
    12 * 12 items/Case
    12 * 12 items/Case
    Specifications
    Environmentally Preferable
  • RANKFE-NMIC-0500
    FE-NMIC-0500
    500 ml
    185 mm
    24/29 NS
    12
    12 * 12 items/Case
    12 * 12 items/Case
    Specifications
    Environmentally Preferable